Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000187.hg.1 |
TC6_apd_hap1000021.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000187.hg.1 |
NM_014596.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000187.hg.1 |
NM_170783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000248.hg.1 |
TC6_mcf_hap5000018.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000248.hg.1 |
NM_014596.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000248.hg.1 |
NM_170783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000332435.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000332435.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000332435.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000332435.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000332435.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000332435.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000332435.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000332435.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000359374.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000359374.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000359374.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000359374.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000359374.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000359374.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000359374.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000376782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000376782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000376782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000376782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000376782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000376782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000376782.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000376785.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000376785.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000376785.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000376785.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000376785.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000376785.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000376785.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000463141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000463141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000463141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000463141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
ENST00000463141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
NM_001278785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
NM_001278785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
NM_001278785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
NM_001278785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
NM_001278785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
NM_001278785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
NM_001278785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
NM_170783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
NM_170783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
NM_170783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
NM_170783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
NM_170783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
NM_170783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
NM_170783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014093.hg.1 |
NM_170783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4000266.hg.1 |
TC6_mann_hap4000024.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4000266.hg.1 |
NM_014596.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4000266.hg.1 |
NM_170783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6000270.hg.1 |
TC6_qbl_hap6000022.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6000270.hg.1 |
NM_014596.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6000270.hg.1 |
NM_170783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3000274.hg.1 |
TC6_dbb_hap3000022.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3000274.hg.1 |
NM_014596.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3000274.hg.1 |
NM_170783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000289.hg.1 |
TC6_cox_hap2000029.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000289.hg.1 |
NM_014596.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000289.hg.1 |
NM_170783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002716.hg.1 |
TC06000330.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002716.hg.1 |
ENST00000332435.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002716.hg.1 |
ENST00000359374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002716.hg.1 |
ENST00000376782.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002716.hg.1 |
ENST00000376785.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002716.hg.1 |
ENST00000463141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002716.hg.1 |
NM_014596.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002716.hg.1 |
NM_170783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000266.hg.1 |
TC6_ssto_hap7000022.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000266.hg.1 |
NM_014596.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000266.hg.1 |
NM_170783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600197868.hg.1 |
ENST00000332435.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600197868.hg.1 |
ENST00000359374.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600197868.hg.1 |
ENST00000376782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600197868.hg.1 |
ENST00000376785.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600197868.hg.1 |
ENST00000463141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600197868.hg.1 |
NM_001278785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600197868.hg.1 |
NM_170783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600197868.hg.1 |
ENST00000332435.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600197868.hg.1 |
ENST00000359374.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600197868.hg.1 |
ENST00000376782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600197868.hg.1 |
ENST00000376785.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600197868.hg.1 |
ENST00000463141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600197868.hg.1 |
NM_001278785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600197868.hg.1 |
NM_170783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600197868.hg.1 |
TC0600014093.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000332435.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000332435.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000332435.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000332435.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000332435.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000332435.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000332435.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000332435.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000359374.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000359374.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000359374.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000359374.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000359374.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000359374.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000359374.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000376782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000376782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000376782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000376782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000376782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000376782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000376782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000376785.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000376785.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000376785.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000376785.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000376785.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000376785.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000376785.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000463141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000463141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000463141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000463141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
ENST00000463141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
NM_001278785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
NM_001278785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
NM_001278785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
NM_001278785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
NM_001278785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
NM_001278785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
NM_001278785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
NM_170783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
NM_170783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
NM_170783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
NM_170783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
NM_170783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
NM_170783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
NM_170783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014093.hg.1 |
NM_170783.3 |
Refseq |