Assay Details
| Gene Symbol: HKDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025130.3 | NP_079406.3 | 1 - 2 | 201 | 61 | |
| XM_011540195.2 | XP_011538497.1 | 1 - 2 | 201 | 61 | ||
| GenBank mRNA | AK095086.1 | - | 1 - 2 | 201 | 61 | |
| AK291024.1 | - | 1 - 2 | 168 | 61 | ||
| BC110504.1 | - | 1 - 2 | 197 | 40028726 | 61 | |
| BC110505.2 | - | 1 - 2 | 200 | 61 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000418.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10004761.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004761.hg.1 | ENST00000395086.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004761.hg.1 | ENST00000395087.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004761.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004761.hg.1 | TC10000418.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004760.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004760.hg.1 | ENST00000395086.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004760.hg.1 | ENST00000395087.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004760.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004760.hg.1 | TC10000418.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000470920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000470920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000470920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000488706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000488706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000488706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153439.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153439.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153439.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153439.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153439.hg.1 | TC1000007890.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000470920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000470920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000470920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000488706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000488706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000488706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153438.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153438.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153438.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153438.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153438.hg.1 | TC1000007890.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000153440.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153440.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153440.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153440.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153440.hg.1 | TC1000007890.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 106473 | - | - |

HKDC1
Genomic Map