Assay Details
| Gene Symbol: HKDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025130.3 | NP_079406.3 | 8 - 9 | 1157 | 87 | |
| XM_011540195.2 | XP_011538497.1 | 8 - 9 | 1157 | 87 | ||
| GenBank mRNA | AK095086.1 | - | 8 - 9 | 1157 | 87 | |
| AK291024.1 | - | 8 - 9 | 1124 | 87 | ||
| AK303408.1 | - | 5 - 6 | 660 | 87 | ||
| BC012337.1 | - | 2 - 3 | 306 | 87 | ||
| BC110504.1 | - | 8 - 9 | 1153 | 40028726 | 87 | |
| BC110505.2 | - | 8 - 9 | 1156 | 87 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000418.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000470920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000470920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000470920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000488706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000488706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | ENST00000488706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004769.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004769.hg.1 | ENST00000395086.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004769.hg.1 | ENST00000395087.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004769.hg.1 | ENST00000486754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004769.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004769.hg.1 | TC10000418.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004770.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004770.hg.1 | ENST00000395086.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004770.hg.1 | ENST00000395087.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004770.hg.1 | ENST00000486754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004770.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10004770.hg.1 | TC10000418.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000153448.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153448.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153448.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153448.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153448.hg.1 | TC1000007890.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000153447.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153447.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153447.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153447.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153447.hg.1 | TC1000007890.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000470920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000470920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000470920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000488706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000488706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | ENST00000488706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007890.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153450.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153450.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153450.hg.1 | ENST00000354624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153450.hg.1 | NM_025130.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153450.hg.1 | TC1000007890.hg.1 | Clariom D TC |

HKDC1
Genomic Map