Assay Details
| Gene Symbol: ASB14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142733.2 | NP_001136205.2 | 10 - 11 | 1911 | 138 | |
| NM_130387.5 | NP_569058.1 | 3 - 4 | 935 | 138 | ||
| GenBank mRNA | AL832472.1 | - | 10 - 11 | 1912 | 138 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001491.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03027111.hg.1 | TC03001491.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027111.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027111.hg.1 | ENST00000438870.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027111.hg.1 | ENST00000487349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027111.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027111.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000515033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000515033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000515033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000515033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | ENST00000515033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011266.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027110.hg.1 | TC03001491.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027110.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027110.hg.1 | ENST00000438870.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027110.hg.1 | ENST00000487349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027110.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027110.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184447.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184447.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184447.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184447.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184447.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184447.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184447.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184447.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184447.hg.1 | TC0300011266.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000515033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000515033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000515033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000515033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | ENST00000515033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011266.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184448.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184448.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184448.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184448.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184448.hg.1 | ENST00000389601.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184448.hg.1 | ENST00000487349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300184448.hg.1 | NM_001142733.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184448.hg.1 | NM_130387.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300184448.hg.1 | TC0300011266.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 192498 | - | - |

ASB14
Genomic Map