Assay Details
| Gene Symbol: TGDS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304430.1 | NP_001291359.1 | 3 - 4 | 430 | 102 | |
| NM_014305.3 | NP_055120.1 | 3 - 4 | 354 | 102 | ||
| NR_130731.1 | 3 - 4 | 354 | 102 | |||
| XM_011521065.2 | XP_011519367.1 | 3 - 4 | 357 | 102 | ||
| XM_011521066.2 | XP_011519368.1 | 3 - 4 | 238 | 102 | ||
| GenBank mRNA | AF048686.1 | - | 3 - 4 | 256 | 102 | |
| AJ006068.1 | - | 3 - 4 | 308 | 102 | ||
| AK313251.1 | - | 3 - 4 | 264 | 102 | ||
| AY544125.1 | - | 3 - 4 | 231 | 102 | ||
| BC005284.1 | - | 3 - 4 | 336 | 102 | ||
| BC033675.1 | - | 3 - 4 | 283 | 5175390 | 102 | |
| BG702561.1 | - | 3 - 4 | 243 | 102 | ||
| BI914642.1 | - | 3 - 4 | 397 | 102 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000793.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000470480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000470480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000470480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13008908.hg.1 | ENST00000261296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13008908.hg.1 | ENST00000498294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13008908.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13008908.hg.1 | TC13000793.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1300163515.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163515.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163515.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163515.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163515.hg.1 | TC1300009492.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300163512.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163512.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163512.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163512.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163512.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163512.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163512.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163512.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163512.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163512.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163512.hg.1 | TC1300009492.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000261296.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000470480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000470480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000470480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | ENST00000498294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_001304430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NM_014305.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130731.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009492.hg.1 | NR_130732.1 | Refseq |

TGDS
Genomic Map