Assay Details
Gene Symbol: RASSF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178169.3 | NP_835463.1 | 2 - 3 | 338 | 134 | |
XM_006719345.3 | XP_006719408.1 | 2 - 3 | 203 | 134 | ||
XM_011538195.2 | XP_011536497.1 | 2 - 3 | 197 | 134 | ||
XM_017019182.1 | XP_016874671.1 | 2 - 3 | 152 | 134 | ||
GenBank mRNA | AY217662.1 | - | 2 - 3 | 338 | 134 | |
BC100949.2 | - | 3 - 4 | 261 | 40004477 | 134 | |
BC100950.1 | - | 3 - 4 | 195 | 40004478 | 134 | |
BC100951.1 | - | 3 - 4 | 261 | 40004481 | 134 | |
BC100952.1 | - | 3 - 4 | 195 | 40004484 | 134 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12000572.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC100950.1,BC100952.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR12007643.hg.1 | ENST00000336061.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007643.hg.1 | ENST00000540088.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007643.hg.1 | ENST00000542104.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007643.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12007643.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12007643.hg.1 | TC12000572.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | ENST00000283172.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | ENST00000283172.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | ENST00000283172.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | ENST00000283172.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | ENST00000283172.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | ENST00000283172.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | ENST00000283172.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | ENST00000283172.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | ENST00000336061.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | ENST00000336061.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | ENST00000336061.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | ENST00000336061.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | ENST00000336061.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | ENST00000336061.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | ENST00000540088.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | ENST00000540088.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | ENST00000540088.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | ENST00000540088.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008005.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12007638.hg.1 | ENST00000540088.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007638.hg.1 | ENST00000541539.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007638.hg.1 | ENST00000542104.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007638.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12007638.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12007638.hg.1 | TC12000572.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12007648.hg.1 | ENST00000336061.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007648.hg.1 | ENST00000541539.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007648.hg.1 | ENST00000542104.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007648.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12007648.hg.1 | TC12000572.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12007640.hg.1 | ENST00000540088.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007640.hg.1 | ENST00000542104.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007640.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12007640.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12007640.hg.1 | TC12000572.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12007647.hg.1 | ENST00000336061.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007647.hg.1 | ENST00000541539.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007647.hg.1 | ENST00000542104.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007647.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12007647.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12007647.hg.1 | TC12000572.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12007644.hg.1 | ENST00000336061.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007644.hg.1 | ENST00000541539.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007644.hg.1 | ENST00000542104.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007644.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12007644.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12007644.hg.1 | TC12000572.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1200158697.hg.1 | ENST00000283172.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158697.hg.1 | ENST00000540088.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158697.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158697.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200158697.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200158697.hg.1 | ENST00000283172.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158697.hg.1 | ENST00000540088.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158697.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158697.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200158697.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200158697.hg.1 | TC1200008005.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200158701.hg.1 | ENST00000283172.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158701.hg.1 | ENST00000336061.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158701.hg.1 | ENST00000540088.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158701.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158701.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200158701.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200158701.hg.1 | ENST00000283172.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158701.hg.1 | ENST00000336061.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158701.hg.1 | ENST00000540088.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158701.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158701.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200158701.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200158701.hg.1 | TC1200008005.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200158705.hg.1 | ENST00000283172.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158705.hg.1 | ENST00000336061.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158705.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158705.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200158705.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200158705.hg.1 | ENST00000283172.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158705.hg.1 | ENST00000336061.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158705.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158705.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200158705.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200158705.hg.1 | TC1200008005.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200158696.hg.1 | ENST00000283172.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158696.hg.1 | ENST00000540088.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158696.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158696.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200158696.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200158696.hg.1 | ENST00000283172.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158696.hg.1 | ENST00000540088.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158696.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158696.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200158696.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200158696.hg.1 | TC1200008005.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200158707.hg.1 | ENST00000336061.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158707.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158707.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200158707.hg.1 | ENST00000336061.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158707.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200158707.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200158707.hg.1 | TC1200008005.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | ENST00000283172.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | ENST00000283172.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | ENST00000283172.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | ENST00000283172.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | ENST00000283172.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | ENST00000283172.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | ENST00000283172.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | ENST00000283172.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | ENST00000336061.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | ENST00000336061.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | ENST00000336061.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | ENST00000336061.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | ENST00000336061.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | ENST00000336061.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | ENST00000540088.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | ENST00000540088.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | ENST00000540088.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | ENST00000540088.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | ENST00000542104.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | NM_178169.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | NR_040718.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008005.hg.1 | NR_040718.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 208499 | - | - |
