Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007199.hg.1 |
TC03000333.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007199.hg.1 |
ENST00000420323.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007199.hg.1 |
NM_015512.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153800.hg.1 |
ENST00000420323.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153800.hg.1 |
ENST00000486752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153800.hg.1 |
ENST00000488988.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153800.hg.1 |
NM_015512.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153800.hg.1 |
ENST00000420323.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153800.hg.1 |
ENST00000486752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153800.hg.1 |
ENST00000488988.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153800.hg.1 |
NM_015512.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153800.hg.1 |
TC0300007474.hg.1 |
Clariom D TC |