Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08008205.hg.1 |
TC08000591.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08008205.hg.1 |
ENST00000220763.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08008205.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000517742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000517742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000517742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000517742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000517742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000517742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000519484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000519484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000519484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000519484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000519900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000519900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000519900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000519900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000519900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000521142.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000521142.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000521142.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000521142.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000522617.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000522617.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000522617.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000522617.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000525310.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000525310.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000525310.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000525310.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000529551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000529551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000529551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000529551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000532528.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000532528.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000532528.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
ENST00000532528.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800157199.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800157199.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800157199.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800157199.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800157199.hg.1 |
TC0800008300.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000517742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000517742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000517742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000517742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000517742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000517742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000519484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000519484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000519484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000519484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000519900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000519900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000519900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000519900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000519900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000521142.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000521142.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000521142.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000521142.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000522617.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000522617.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000522617.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000522617.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000525310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000525310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000525310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000525310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000529551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000529551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000529551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000529551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000532528.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000532528.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000532528.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
ENST00000532528.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008300.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800157197.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800157197.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800157197.hg.1 |
ENST00000220763.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800157197.hg.1 |
NM_016134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800157197.hg.1 |
TC0800008300.hg.1 |
Clariom D TC |