Assay Details
Gene Symbol: CPQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016134.3 | NP_057218.1 | 4 - 5 | 1052 | 91 | |
GenBank mRNA | AF107834.1 | - | 3 - 4 | 895 | 91 | |
AF119386.1 | - | 3 - 4 | 866 | 91 | ||
AK075132.1 | - | 4 - 5 | 970 | 91 | ||
AK296013.1 | - | 4 - 5 | 672 | 91 | ||
AK304008.1 | - | 4 - 5 | 448 | 91 | ||
AK315447.1 | - | 4 - 5 | 969 | 91 | ||
BC020689.1 | - | 4 - 5 | 1008 | 4251802 | 91 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08000591.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR08008206.hg.1 | TC08000591.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08008206.hg.1 | ENST00000220763.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08008206.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000519484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000519484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000519484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000519484.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000519900.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000519900.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000519900.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000519900.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000519900.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000521142.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000521142.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000521142.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000521142.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000522617.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000522617.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000522617.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000522617.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000525310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000525310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000525310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000525310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000529551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000529551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000529551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000529551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000532528.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000532528.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000532528.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | ENST00000532528.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000517742.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000519484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000519484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000519484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000519484.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000519900.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000519900.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000519900.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000519900.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000519900.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000521142.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000521142.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000521142.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000521142.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000522617.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000522617.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000522617.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000522617.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000525310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000525310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000525310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000525310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000529551.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000529551.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000529551.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000529551.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000532528.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000532528.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000532528.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | ENST00000532528.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008300.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800157200.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157200.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800157200.hg.1 | ENST00000220763.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800157200.hg.1 | NM_016134.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800157200.hg.1 | TC0800008300.hg.1 | Clariom D TC |
