Assay Details
| Gene Symbol: KCNQ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011528810.1 | XP_011527112.1 | 15 - 16 | 1948 | 77 | |
| XM_011528811.1 | XP_011527113.1 | 14 - 15 | 1918 | 77 | ||
| XM_017027841.1 | XP_016883330.1 | 15 - 16 | 1945 | 77 | ||
| XM_017027843.1 | XP_016883332.1 | 15 - 16 | 1841 | 77 | ||
| XM_017027845.1 | XP_016883334.1 | 5 - 6 | 1388 | 77 | ||
More Information
Set Membership: |
|

KCNQ2
Genomic Map