Assay Details
Gene Symbol: LRGUK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144648.1 | NP_653249.1 | 15 - 16 | 1859 | 72 | |
GenBank mRNA | AK057348.1 | - | 15 - 16 | 1859 | 72 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07000831.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR07013088.hg.1 | TC07000831.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07013088.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07013088.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | ENST00000473068.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | ENST00000473068.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | ENST00000473068.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | ENST00000473068.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07013087.hg.1 | TC07000831.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07013087.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07013087.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700165553.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700165553.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700165553.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700165553.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700165553.hg.1 | TC0700009222.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700165552.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700165552.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700165552.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700165552.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700165552.hg.1 | TC0700009222.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | ENST00000285928.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | ENST00000473068.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | ENST00000473068.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | ENST00000473068.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | ENST00000473068.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | NM_144648.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009222.hg.1 | NM_144648.2 | Refseq |
