Assay Details
| Gene Symbol: COLQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005677.3 | NP_005668.2 | 8 - 9 | 680 | 77 | |
| NM_080538.2 | NP_536799.1 | 8 - 9 | 809 | 77 | ||
| NM_080539.3 | NP_536800.2 | 7 - 8 | 578 | 77 | ||
| GenBank mRNA | AF057036.1 | - | 8 - 9 | 680 | 77 | |
| AJ225895.1 | - | 8 - 9 | 673 | 77 | ||
| AK128401.1 | - | 8 - 9 | 809 | 77 | ||
| AK289688.1 | - | 8 - 9 | 699 | 77 | ||
| AY150334.1 | - | 8 - 9 | 657 | 77 | ||
| AY150335.1 | - | 8 - 9 | 657 | 77 | ||
| AY150336.1 | - | 8 - 9 | 657 | 77 | ||
| AY150337.1 | - | 8 - 9 | 657 | 77 | ||
| AY150338.1 | - | 8 - 9 | 657 | 77 | ||
| AY150339.1 | - | 8 - 9 | 657 | 77 | ||
| BC074828.2 | - | 8 - 9 | 584 | 30915542 | 77 | |
| BC074829.2 | - | 8 - 9 | 584 | 30915325 | 77 | |
| BX500613.1 | - | 7 - 8 | 565 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03003335.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383781.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383781.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383781.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383781.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383781.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383781.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383781.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383781.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383781.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383781.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383781.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383781.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383781.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383781.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383781.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383781.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383781.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383781.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383781.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383781.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383781.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383781.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383781.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383781.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383781.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383786.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383786.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383786.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383786.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383786.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383786.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383786.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383786.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383786.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383786.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383786.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383786.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383786.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383786.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383786.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383786.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383786.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383786.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383786.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383786.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383786.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000383788.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000603469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000603469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000603469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000603752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000603752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000603808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000603808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000603808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000603808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000603808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000603808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000603808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000603808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000603808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000603808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000603808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000603808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000603808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000603808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000603808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000603808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000603808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000603808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000603808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000603808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000603808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000603808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000603808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000604401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000604401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000604401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000604401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000605797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000605797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000605797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000605797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000605797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000605797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000605797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000605797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000605797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000605797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | ENST00000605797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_005677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_005677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_005677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_005677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_005677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_005677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_005677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_005677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_005677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_005677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_005677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_005677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_005677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_005677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_005677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_005677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_005677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_005677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_005677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_005677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_005677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_005677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_005677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_005677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_005677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_005677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_005677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_005677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080538.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080538.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080538.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080538.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080538.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080538.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080538.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080538.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080538.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080538.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080538.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080538.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080538.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080538.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080538.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080538.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080538.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080538.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080538.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080538.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080538.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080538.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080538.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080538.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080538.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080538.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080538.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080538.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080539.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080539.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080539.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080539.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080539.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080539.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080539.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080539.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080539.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080539.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080539.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080539.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080539.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080539.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080539.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080539.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080539.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080539.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080539.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080539.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080539.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080539.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080539.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080539.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080539.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080539.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013947.hg.1 | NM_080539.3 | Refseq |

COLQ
Genomic Map