Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01012027.hg.1 |
TC01000752.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01012027.hg.1 |
ENST00000035383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01012027.hg.1 |
ENST00000310961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01012027.hg.1 |
ENST00000370957.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01012027.hg.1 |
ENST00000415775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01012027.hg.1 |
NM_020794.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100161772.hg.1 |
ENST00000035383.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100161772.hg.1 |
ENST00000310961.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100161772.hg.1 |
ENST00000415775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100161772.hg.1 |
NM_020794.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100161772.hg.1 |
ENST00000035383.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100161772.hg.1 |
ENST00000310961.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100161772.hg.1 |
ENST00000415775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100161772.hg.1 |
NM_020794.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100161772.hg.1 |
TC0100008686.hg.1 |
Clariom D TC |