Assay Details
| Gene Symbol: PITX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204397.1 | NP_001191326.1 | 4 - 5 | 1871 | 122 | |
| NM_001204398.1 | NP_001191327.1 | 2 - 3 | 263 | 122 | ||
| NM_153426.2 | NP_700475.1 | 3 - 4 | 1637 | 122 | ||
| GenBank mRNA | AF048721.1 | - | 2 - 3 | 298 | 122 | |
| AK127829.1 | - | 4 - 5 | 1709 | 122 | ||
| BP372081.1 | - | 2 - 3 | 263 | 122 | ||
More Information
Set Membership: |
|

PITX2
Genomic Map