Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014868.hg.1 |
ENST00000261978.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014868.hg.1 |
ENST00000556690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014868.hg.1 |
NM_000428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014868.hg.1 |
TC14001308.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400171412.hg.1 |
ENST00000261978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400171412.hg.1 |
ENST00000553939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400171412.hg.1 |
ENST00000556206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400171412.hg.1 |
ENST00000556690.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400171412.hg.1 |
NM_000428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400171412.hg.1 |
ENST00000261978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400171412.hg.1 |
ENST00000553939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400171412.hg.1 |
ENST00000556206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400171412.hg.1 |
ENST00000556690.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400171412.hg.1 |
NM_000428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400171412.hg.1 |
TC1400009698.hg.1 |
Clariom D TC |