Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039177.hg.1 |
TC02002482.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039177.hg.1 |
ENST00000328032.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039177.hg.1 |
ENST00000332142.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039177.hg.1 |
ENST00000477019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039177.hg.1 |
NM_033272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039177.hg.1 |
NM_173162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200207630.hg.1 |
ENST00000328032.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207630.hg.1 |
ENST00000332142.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207630.hg.1 |
ENST00000477019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207630.hg.1 |
ENST00000618399.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207630.hg.1 |
ENST00000621889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207630.hg.1 |
NM_033272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200207630.hg.1 |
NM_173162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200207630.hg.1 |
ENST00000328032.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207630.hg.1 |
ENST00000332142.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207630.hg.1 |
ENST00000477019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207630.hg.1 |
ENST00000618399.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207630.hg.1 |
ENST00000621889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207630.hg.1 |
NM_033272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200207630.hg.1 |
NM_173162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200207630.hg.1 |
TC0200014775.hg.1 |
Clariom D TC |