Assay Details
Gene Symbol: KCNH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033272.3 | NP_150375.2 | 8 - 9 | 2182 | 135 | |
NM_173162.2 | NP_775185.1 | 7 - 8 | 2161 | 135 | ||
XM_011512109.2 | XP_011510411.1 | 9 - 10 | 2096 | 135 | ||
XM_017005218.1 | XP_016860707.1 | 9 - 10 | 2096 | 135 | ||
XM_017005219.1 | XP_016860708.1 | 8 - 9 | 2072 | 135 | ||
XM_017005220.1 | XP_016860709.1 | 7 - 8 | 2051 | 135 | ||
GenBank mRNA | AF032897.1 | - | 8 - 9 | 2053 | 135 | |
BC035815.1 | - | 7 - 8 | 2144 | 5769938 | 135 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02002482.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02039169.hg.1 | TC02002482.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02039169.hg.1 | ENST00000332142.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02039169.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02039170.hg.1 | TC02002482.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02039170.hg.1 | ENST00000328032.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02039170.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02039171.hg.1 | TC02002482.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02039171.hg.1 | ENST00000328032.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02039171.hg.1 | ENST00000332142.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02039171.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02039171.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200207621.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200207621.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200207621.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200207621.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200207621.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200207621.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200207621.hg.1 | TC0200014775.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200207623.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200207623.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200207623.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200207623.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200207623.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200207623.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200207623.hg.1 | TC0200014775.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200207624.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200207624.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200207624.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200207624.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200207624.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200207624.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200207624.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200207624.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200207624.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200207624.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200207624.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200207624.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200207624.hg.1 | TC0200014775.hg.1 | Clariom D TC |
