Assay Details
Gene Symbol: MIF4GD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011525051.2 | XP_011523353.1 | 1 - 2 | 301 | 63 | |
XM_011525052.2 | XP_011523354.1 | 1 - 2 | 301 | 63 | ||
XM_011525053.2 | XP_011523355.1 | 1 - 2 | 301 | 63 | ||
XM_011525054.2 | XP_011523356.1 | 1 - 2 | 301 | 63 | ||
XM_011525055.2 | XP_011523357.1 | 1 - 2 | 301 | 63 | ||
XM_017024888.1 | XP_016880377.1 | 1 - 2 | 301 | 63 | ||
XM_017024892.1 | XP_016880381.1 | 1 - 2 | 301 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
