Assay Details
| Gene Symbol: FUZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011527340.1 | XP_011525642.1 | 1 - 2 | 154 | 56 | |
| XM_017027319.1 | XP_016882808.1 | 1 - 2 | 154 | 56 | ||
| XM_017027322.1 | XP_016882811.1 | 1 - 2 | 154 | 56 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011527340.1, XM_017027319.1, XM_017027322.1 transcripts.
Set Membership: |
|

FUZ
Genomic Map