Assay Details
Gene Symbol: PPM1K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152542.4 | NP_689755.3 | 4 - 5 | 1196 | 86 | |
XM_006714111.3 | XP_006714174.1 | 4 - 5 | 1257 | 86 | ||
XM_017007803.1 | XP_016863292.1 | 4 - 5 | 844 | 86 | ||
XM_017007804.1 | XP_016863293.1 | 5 - 6 | 1384 | 86 | ||
XM_017007805.1 | XP_016863294.1 | 6 - 7 | 1384 | 86 | ||
XM_017007806.1 | XP_016863295.1 | 4 - 5 | 717 | 86 | ||
XM_017007807.1 | XP_016863296.1 | 3 - 4 | 435 | 86 | ||
GenBank mRNA | AK054678.1 | - | 4 - 5 | 924 | 86 | |
AK314417.1 | - | 4 - 5 | 921 | 86 | ||
AL834271.1 | - | 4 - 5 | 915 | 86 | ||
AX766363.1 | - | 3 - 4 | 711 | 86 | ||
AY157615.1 | - | 4 - 5 | 924 | 86 | ||
AY435431.1 | - | 4 - 5 | 1101 | 86 | ||
AY994097.1 | - | 4 - 5 | 932 | 86 | ||
BC037552.1 | - | 4 - 5 | 960 | 5227317 | 86 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04001373.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR04019203.hg.1 | TC04001373.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04019203.hg.1 | ENST00000295908.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04019203.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000363599.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000505022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000505022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000505022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000505022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000509340.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000509340.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000509340.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000511506.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000511506.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000511506.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000363599.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000505022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000505022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000505022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000505022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000509340.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000509340.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000509340.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000511506.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000511506.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000511506.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400177171.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400177171.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400177171.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400177171.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400177171.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400177171.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400177171.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400177171.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400177171.hg.1 | TC0400011283.hg.1 | Clariom D TC |
