Assay Details
Gene Symbol: SDF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF153686.1 | - | 4 - 5 | 871 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000360001.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000360001.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000360001.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000360001.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000360001.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000360001.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000360001.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000360001.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000360001.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000360001.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000360001.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000360001.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000360001.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000360001.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000403997.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000403997.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000403997.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000403997.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000403997.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000403997.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000459994.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000459994.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000459994.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000459994.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000459994.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000459994.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000459994.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000478938.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000478938.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000478938.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000478938.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000478938.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000478938.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000478938.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000478938.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000494748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000494748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000494748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000494748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000494748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000494748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000494748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | ENST00000494748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000263741.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000360001.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000360001.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000360001.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000360001.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000360001.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000360001.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000360001.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000360001.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000360001.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000360001.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000360001.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000360001.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000360001.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000360001.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000403997.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000403997.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000403997.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000403997.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000403997.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000403997.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000459994.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000459994.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000459994.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000459994.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000459994.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000459994.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000459994.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000465727.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000478938.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000478938.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000478938.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000478938.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000478938.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000478938.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000478938.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000478938.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000494748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000494748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000494748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000494748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000494748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000494748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000494748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | ENST00000494748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016176.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016547.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012454.hg.1 | NM_016547.2 | Refseq |
