| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06024544.hg.1 |
TC06001980.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06024544.hg.1 |
ENST00000369110.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06024544.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06024545.hg.1 |
TC06001980.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06024545.hg.1 |
ENST00000369110.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06024545.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
ENST00000448705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
ENST00000448705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600187687.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600187687.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600187687.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600187687.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600187687.hg.1 |
TC0600012713.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600187688.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600187688.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600187688.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600187688.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600187688.hg.1 |
TC0600012713.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
ENST00000369110.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
ENST00000448705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
ENST00000448705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012713.hg.1 |
NM_002726.4 |
Refseq |