Assay Details
| Gene Symbol: BPGM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199186.2 | NP_954655.1 | 1 - 2 | 157 | 125 | |
| GenBank mRNA | DB049554.1 | - | 1 - 2 | 104 | 125 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000836.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_199186.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
carbohydrate metabolic process
glucose metabolic process
gluconeogenesis
glycolytic process
respiratory gaseous exchange
dephosphorylation
regulation of pentose-phosphate shunt
bisphosphoglycerate mutase activity
bisphosphoglycerate 2-phosphatase activity
2,3-bisphosphoglycerate-dependent phosphoglycerate mutase activity
carbohydrate metabolic process
glucose metabolic process
gluconeogenesis
glycolytic process
respiratory gaseous exchange
dephosphorylation
regulation of pentose-phosphate shunt
bisphosphoglycerate mutase activity
bisphosphoglycerate 2-phosphatase activity
2,3-bisphosphoglycerate-dependent phosphoglycerate mutase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | ENST00000344924.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | ENST00000344924.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | ENST00000344924.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | ENST00000344924.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | ENST00000344924.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | ENST00000344924.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | ENST00000393132.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | ENST00000393132.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | ENST00000393132.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | ENST00000393132.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | ENST00000393132.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | ENST00000393132.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | ENST00000393132.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | ENST00000418040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | ENST00000418040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | ENST00000418040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | ENST00000418040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | ENST00000418040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | ENST00000418040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | ENST00000418040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | ENST00000443095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | ENST00000443095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | ENST00000443095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | NM_001293085.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | NM_001293085.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | NM_001293085.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | NM_001293085.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | NM_001293085.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | NM_001293085.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | NM_001293085.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | NM_001293085.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | NM_001724.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | NM_001724.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | NM_001724.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | NM_001724.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | NM_001724.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | NM_001724.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | NM_001724.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | NM_199186.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | NM_199186.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | NM_199186.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | NM_199186.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | NM_199186.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | NM_199186.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | NM_199186.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009232.hg.1 | NM_199186.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07013141.hg.1 | TC07000836.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07013141.hg.1 | ENST00000393132.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07013141.hg.1 | NM_199186.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | ENST00000344924.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | ENST00000344924.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | ENST00000344924.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | ENST00000344924.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | ENST00000344924.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | ENST00000344924.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | ENST00000393132.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | ENST00000393132.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | ENST00000393132.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | ENST00000393132.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | ENST00000393132.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | ENST00000393132.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | ENST00000393132.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | ENST00000418040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | ENST00000418040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | ENST00000418040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | ENST00000418040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | ENST00000418040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | ENST00000418040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | ENST00000418040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | ENST00000443095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | ENST00000443095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | ENST00000443095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | NM_001293085.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | NM_001293085.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | NM_001293085.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | NM_001293085.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | NM_001293085.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | NM_001293085.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | NM_001293085.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | NM_001293085.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | NM_001724.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | NM_001724.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | NM_001724.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | NM_001724.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | NM_001724.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | NM_001724.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | NM_001724.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | NM_199186.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | NM_199186.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | NM_199186.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | NM_199186.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | NM_199186.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | NM_199186.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | NM_199186.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009232.hg.1 | NM_199186.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700165623.hg.1 | ENST00000393132.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700165623.hg.1 | NM_199186.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700165623.hg.1 | ENST00000393132.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700165623.hg.1 | NM_199186.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700165623.hg.1 | TC0700009232.hg.1 | Clariom D TC |

BPGM
Genomic Map