Assay Details
Gene Symbol: NR2F1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_109825.1 | 6 - 7 | 1328 | 143 | ||
GenBank mRNA | BC044619.1 | - | 6 - 7 | 804 | 143 | |
BC048999.1 | - | 6 - 7 | 804 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
