Assay Details
Gene Symbol: C19orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006722885.3 | XP_006722948.2 | 2 - 3 | 460 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000585755.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000585755.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000585755.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586500.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586500.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586500.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586500.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000589393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000589393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000589393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000589393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591439.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591439.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591439.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000585755.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000585755.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000585755.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586500.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586500.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586500.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586500.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000589393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000589393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000589393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000589393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591439.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591439.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591439.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
