Assay Details
| Gene Symbol: BCCIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016567.3 | NP_057651.1 | 5 - 6 | 622 | 115 | |
| NM_078468.2 | NP_510868.1 | 5 - 6 | 622 | 115 | ||
| NM_078469.2 | NP_510869.1 | 5 - 6 | 622 | 115 | ||
| GenBank mRNA | AB040450.1 | - | 5 - 6 | 611 | 115 | |
| AB040451.1 | - | 5 - 6 | 611 | 115 | ||
| AK055691.1 | - | 5 - 6 | 611 | 115 | ||
| AK300767.1 | - | 5 - 6 | 602 | 115 | ||
| AK304529.1 | - | 4 - 5 | 521 | 115 | ||
| AL834458.1 | - | 5 - 6 | 600 | 115 | ||
| AY064248.1 | - | 5 - 6 | 622 | 115 | ||
| AY064249.1 | - | 5 - 6 | 622 | 115 | ||
| BC009771.1 | - | 5 - 6 | 615 | 4277458 | 115 | |
| BM905542.1 | - | 645 | 115 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000915.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000278100.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000278100.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000278100.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000278100.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000278100.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000278100.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000278100.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000278100.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000278100.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000278100.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000299130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000299130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000299130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000299130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000299130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000299130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000299130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000299130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000299130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000299130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000299130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000368759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000368759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000368759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000368759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000368759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000368759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000368759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000368759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000368759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000368759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000368759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000463330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000463330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000463330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000463330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000463330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000463330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000478798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000478798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000478798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000478798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | ENST00000478798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_016567.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_016567.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_016567.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_016567.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_016567.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_016567.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_016567.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_016567.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_016567.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_016567.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_016567.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_078468.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_078468.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_078468.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_078468.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_078468.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_078468.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_078468.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_078468.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_078468.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_078468.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_078469.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_078469.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_078469.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_078469.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_078469.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_078469.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_078469.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_078469.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_078469.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_078469.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009244.hg.1 | NM_078469.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011205.hg.1 | ENST00000278100.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011205.hg.1 | ENST00000299130.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011205.hg.1 | ENST00000368759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011205.hg.1 | ENST00000392718.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011205.hg.1 | ENST00000429863.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011205.hg.1 | NM_016567.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011205.hg.1 | NM_078468.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011205.hg.1 | NM_078469.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011205.hg.1 | TC10000915.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011203.hg.1 | ENST00000278100.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011203.hg.1 | ENST00000299130.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011203.hg.1 | ENST00000368759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011203.hg.1 | ENST00000392718.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011203.hg.1 | ENST00000429863.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011203.hg.1 | NM_016567.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011203.hg.1 | NM_078468.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011203.hg.1 | NM_078469.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011203.hg.1 | TC10000915.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000163414.hg.1 | ENST00000278100.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000163414.hg.1 | ENST00000299130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000163414.hg.1 | ENST00000368759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000163414.hg.1 | NM_016567.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000163414.hg.1 | NM_078468.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000163414.hg.1 | NM_078469.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000163414.hg.1 | ENST00000278100.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000163414.hg.1 | ENST00000299130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000163414.hg.1 | ENST00000368759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000163414.hg.1 | NM_016567.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000163414.hg.1 | NM_078468.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000163414.hg.1 | NM_078469.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000163414.hg.1 | TC1000009244.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000163412.hg.1 | ENST00000278100.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000163412.hg.1 | ENST00000299130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000163412.hg.1 | ENST00000368759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000163412.hg.1 | ENST00000463330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000163412.hg.1 | NM_016567.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000163412.hg.1 | NM_078468.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000163412.hg.1 | NM_078469.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000163412.hg.1 | ENST00000278100.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000163412.hg.1 | ENST00000299130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000163412.hg.1 | ENST00000368759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000163412.hg.1 | ENST00000463330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000163412.hg.1 | NM_016567.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000163412.hg.1 | NM_078468.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000163412.hg.1 | NM_078469.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000163412.hg.1 | TC1000009244.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000278100.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000278100.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000278100.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000278100.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000278100.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000278100.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000278100.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000278100.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000278100.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000278100.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000299130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000299130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000299130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000299130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000299130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000299130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000299130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000299130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000299130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000299130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000299130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000368759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000368759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000368759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000368759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000368759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000368759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000368759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000368759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000368759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000368759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000368759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000463330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000463330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000463330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000463330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000463330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000463330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000478798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000478798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000478798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000478798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | ENST00000478798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_016567.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_016567.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_016567.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_016567.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_016567.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_016567.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_016567.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_016567.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_016567.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_016567.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_016567.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_078468.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_078468.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_078468.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_078468.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_078468.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_078468.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_078468.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_078468.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_078468.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_078468.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_078469.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_078469.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_078469.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_078469.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_078469.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_078469.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_078469.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_078469.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_078469.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_078469.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009244.hg.1 | NM_078469.2 | Refseq |

BCCIP
Genomic Map