Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021616.hg.1 |
TC02001377.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021616.hg.1 |
ENST00000340126.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021616.hg.1 |
ENST00000414648.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021616.hg.1 |
NM_001080391.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200179110.hg.1 |
ENST00000340126.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179110.hg.1 |
ENST00000431952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179110.hg.1 |
ENST00000488180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179110.hg.1 |
ENST00000494901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179110.hg.1 |
NM_001080391.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200179110.hg.1 |
ENST00000340126.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179110.hg.1 |
ENST00000431952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179110.hg.1 |
ENST00000488180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179110.hg.1 |
ENST00000494901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179110.hg.1 |
NM_001080391.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200179110.hg.1 |
TC0200011023.hg.1 |
Clariom D TC |