Assay Details
| Gene Symbol: GPRC5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006721051.2 | XP_006721114.1 | 1 - 2 | 177 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006721051.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
G-protein coupled receptor signaling pathway
locomotory behavior
positive regulation of neuron projection development
activation of protein kinase activity
glucose homeostasis
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of neuron differentiation
positive regulation of inflammatory response
positive regulation of macrophage cytokine production
positive regulation of protein tyrosine kinase activity
positive regulation of canonical Wnt signaling pathway
G-protein coupled receptor binding
G-protein coupled receptor activity
protein kinase binding
protein kinase activator activity
G-protein coupled receptor signaling pathway
locomotory behavior
positive regulation of neuron projection development
activation of protein kinase activity
glucose homeostasis
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of neuron differentiation
positive regulation of inflammatory response
positive regulation of macrophage cytokine production
positive regulation of protein tyrosine kinase activity
positive regulation of canonical Wnt signaling pathway
G-protein coupled receptor binding
G-protein coupled receptor activity
protein kinase binding
protein kinase activator activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000535671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000535671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000535671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000535671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000535671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000535671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000535671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000562348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000562348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000562348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000562348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000562348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000562469.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000562469.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000562469.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000562469.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000562469.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000562469.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000562469.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000564449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000564449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000564449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000566822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000566822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000566822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000566822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000566822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000566822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000568214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000568214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000568214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000568214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000569479.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000569479.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000569479.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000569479.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000569479.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000569479.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000569479.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000569479.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000569479.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000569479.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000569847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000569847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000569847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000569847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000569847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000569847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000569847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000569847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000569847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000569847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000569847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000570142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000570142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000570142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000570142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | ENST00000570142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | NM_001304771.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | NM_001304771.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | NM_001304771.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | NM_001304771.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | NM_001304771.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | NM_001304771.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | NM_001304771.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | NM_001304771.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | NM_001304771.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | NM_001304771.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | NM_001304771.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | NM_016235.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | NM_016235.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | NM_016235.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | NM_016235.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | NM_016235.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | NM_016235.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | NM_016235.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | NM_016235.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | NM_016235.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | NM_016235.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | NM_016235.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011496.hg.1 | NM_016235.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000535671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000535671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000535671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000535671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000535671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000535671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000535671.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000562348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000562348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000562348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000562348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000562348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000562469.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000562469.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000562469.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000562469.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000562469.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000562469.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000562469.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000564449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000564449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000564449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000566822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000566822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000566822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000566822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000566822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000566822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000568214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000568214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000568214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000568214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000569479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000569479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000569479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000569479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000569479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000569479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000569479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000569479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000569479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000569479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000569847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000569847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000569847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000569847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000569847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000569847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000569847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000569847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000569847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000569847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000569847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000570142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000570142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000570142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000570142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | ENST00000570142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | NM_001304771.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | NM_001304771.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | NM_001304771.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | NM_001304771.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | NM_001304771.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | NM_001304771.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | NM_001304771.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | NM_001304771.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | NM_001304771.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | NM_001304771.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | NM_001304771.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | NM_016235.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | NM_016235.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | NM_016235.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | NM_016235.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | NM_016235.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | NM_016235.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | NM_016235.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | NM_016235.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | NM_016235.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | NM_016235.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | NM_016235.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011496.hg.1 | NM_016235.2 | Refseq |

GPRC5B
Genomic Map