Assay Details
Gene Symbol: PLAU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011539866.2 | XP_011538168.1 | 1 - 2 | 107 | 143 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011539866.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
proteolysis
chemotaxis
signal transduction
spermatogenesis
embryo implantation
blood coagulation
positive regulation of cell proliferation
regulation of receptor activity
response to activity
smooth muscle cell migration
regulation of smooth muscle cell migration
positive regulation of smooth muscle cell migration
positive regulation of cell migration
regulation of cell adhesion mediated by integrin
cellular response to hepatocyte growth factor stimulus
fibrinolysis
skeletal muscle tissue regeneration
response to hyperoxia
positive regulation of ovulation
regulation of wound healing
neuron death
cellular response to lipopolysaccharide
cellular response to glucose stimulus
cellular response to hypoxia
cellular response to fluid shear stress
cellular response to staurosporine
regulation of smooth muscle cell-matrix adhesion
regulation of hepatocyte proliferation
positive regulation of reactive oxygen species metabolic process
serine-type endopeptidase activity
protein binding
angiogenesis
proteolysis
chemotaxis
signal transduction
spermatogenesis
embryo implantation
blood coagulation
positive regulation of cell proliferation
regulation of receptor activity
response to activity
smooth muscle cell migration
regulation of smooth muscle cell migration
positive regulation of smooth muscle cell migration
positive regulation of cell migration
regulation of cell adhesion mediated by integrin
cellular response to hepatocyte growth factor stimulus
fibrinolysis
skeletal muscle tissue regeneration
response to hyperoxia
positive regulation of ovulation
regulation of wound healing
neuron death
cellular response to lipopolysaccharide
cellular response to glucose stimulus
cellular response to hypoxia
cellular response to fluid shear stress
cellular response to staurosporine
regulation of smooth muscle cell-matrix adhesion
regulation of hepatocyte proliferation
positive regulation of reactive oxygen species metabolic process
serine-type endopeptidase activity
protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000372764.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000372764.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000372764.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000372764.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000372764.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000372764.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000372764.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000372764.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000372764.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000372764.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000372764.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000372764.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000372764.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000481390.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000481390.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000481390.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000481390.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000481390.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000494287.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000494287.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000494287.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000494287.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000494287.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000494287.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000494287.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | ENST00000496926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | NM_002658.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | NM_002658.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | NM_002658.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | NM_002658.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | NM_002658.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | NM_002658.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | NM_002658.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | NM_002658.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | NM_002658.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | NM_002658.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | NM_002658.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | NM_002658.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | NM_002658.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008054.hg.1 | NM_002658.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000372764.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000372764.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000372764.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000372764.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000372764.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000372764.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000372764.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000372764.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000372764.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000372764.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000372764.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000372764.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000372764.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000446342.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000481390.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000481390.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000481390.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000481390.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000481390.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000494287.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000494287.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000494287.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000494287.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000494287.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000494287.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000494287.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | ENST00000496926.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | NM_001145031.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | NM_002658.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | NM_002658.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | NM_002658.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | NM_002658.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | NM_002658.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | NM_002658.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | NM_002658.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | NM_002658.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | NM_002658.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | NM_002658.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | NM_002658.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | NM_002658.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | NM_002658.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008054.hg.1 | NM_002658.4 | Refseq |
