Assay Details
| Gene Symbol: RALA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005402.3 | NP_005393.2 | 3 - 4 | 630 | 137 | |
| XM_006715762.2 | XP_006715825.1 | 3 - 4 | 422 | 137 | ||
| XM_011515466.1 | XP_011513768.1 | 4 - 5 | 674 | 137 | ||
| GenBank mRNA | AF493910.1 | - | 2 - 3 | 329 | 137 | |
| AK308109.1 | - | 3 - 4 | 483 | 137 | ||
| BC039858.1 | - | 3 - 4 | 576 | 5495399 | 137 | |
| M29893.1 | - | 2 - 3 | 329 | 137 | ||
| X15014.1 | - | 2 - 3 | 320 | 137 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000250.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cytokinesis
neural tube closure
exocytosis
chemotaxis
signal transduction
Ras protein signal transduction
viral process
regulation of exocytosis
actin cytoskeleton reorganization
positive regulation of filopodium assembly
membrane raft localization
membrane organization
GTPase activity
protein binding
GTP binding
myosin binding
GDP binding
ubiquitin protein ligase binding
Edg-2 lysophosphatidic acid receptor binding
ATPase binding
cytokinesis
neural tube closure
exocytosis
chemotaxis
signal transduction
Ras protein signal transduction
viral process
regulation of exocytosis
actin cytoskeleton reorganization
positive regulation of filopodium assembly
membrane raft localization
membrane organization
GTPase activity
protein binding
GTP binding
myosin binding
GDP binding
ubiquitin protein ligase binding
Edg-2 lysophosphatidic acid receptor binding
ATPase binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07003841.hg.1 | TC07000250.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003841.hg.1 | ENST00000005257.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003841.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003844.hg.1 | TC07000250.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003844.hg.1 | ENST00000005257.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003844.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003844.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003837.hg.1 | TC07000250.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003837.hg.1 | ENST00000005257.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003837.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003837.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003838.hg.1 | TC07000250.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003838.hg.1 | ENST00000005257.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003838.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003838.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003840.hg.1 | TC07000250.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003840.hg.1 | ENST00000005257.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003840.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006850.hg.1 | ENST00000447806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006850.hg.1 | ENST00000447806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003842.hg.1 | TC07000250.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003842.hg.1 | ENST00000005257.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003842.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000436179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000436179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000436179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000436179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000466491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000466491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000466491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700150442.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150442.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150442.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700150442.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150442.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150442.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700150442.hg.1 | TC0700007271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700006850.hg.1 | ENST00000447806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006850.hg.1 | ENST00000447806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150447.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150447.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150447.hg.1 | ENST00000436179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150447.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700150447.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150447.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150447.hg.1 | ENST00000436179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150447.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700150447.hg.1 | TC0700007271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000436179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000436179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000436179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000436179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000466491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000466491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000466491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700150449.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150449.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150449.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150449.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700150449.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150449.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150449.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150449.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700150449.hg.1 | TC0700007271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700150444.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150444.hg.1 | ENST00000436179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150444.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700150444.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150444.hg.1 | ENST00000436179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150444.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700150444.hg.1 | TC0700007271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700150441.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150441.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150441.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700150441.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150441.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150441.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700150441.hg.1 | TC0700007271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700150446.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150446.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150446.hg.1 | ENST00000436179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150446.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700150446.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150446.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150446.hg.1 | ENST00000436179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700150446.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700150446.hg.1 | TC0700007271.hg.1 | Clariom D TC |

RALA
Genomic Map