Assay Details
| Gene Symbol: MRPL24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024540.3 | NP_078816.2 | 1 - 2 | 75 | 63 | |
| GenBank mRNA | BC012440.1 | - | 1 - 2 | 57 | 3863359 | 63 |
| BG706700.1 | - | 1 - 2 | 80 | 63 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003352.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC012440.1,NM_024540.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01052349.hg.1 | TC01003352.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01052349.hg.1 | ENST00000361531.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01052349.hg.1 | ENST00000368211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01052349.hg.1 | NM_024540.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01052349.hg.1 | NM_145729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01052353.hg.1 | TC01003352.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01052353.hg.1 | ENST00000361531.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01052353.hg.1 | ENST00000368211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01052353.hg.1 | NM_024540.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01052353.hg.1 | NM_145729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000361531.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000361531.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000361531.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000361531.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000361531.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000361531.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000361531.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000361531.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000361531.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000368211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000368211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000368211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000368211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000368211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000368211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000368211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000368211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000368211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000412846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000412846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000412846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000412846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000412846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000412846.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000420938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000420938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000420938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000434558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000434558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000434558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000434558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000434558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000434558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000478899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000478899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000478899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000478899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | ENST00000478899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | NM_024540.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | NM_024540.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | NM_024540.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | NM_024540.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | NM_024540.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | NM_024540.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | NM_024540.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | NM_024540.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | NM_024540.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | NM_145729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | NM_145729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | NM_145729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | NM_145729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | NM_145729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | NM_145729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | NM_145729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | NM_145729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | NM_145729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016021.hg.1 | NM_145729.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221572.hg.1 | ENST00000361531.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221572.hg.1 | ENST00000368211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221572.hg.1 | ENST00000412846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221572.hg.1 | ENST00000420938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221572.hg.1 | ENST00000434558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221572.hg.1 | NM_024540.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221572.hg.1 | NM_145729.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221572.hg.1 | ENST00000361531.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221572.hg.1 | ENST00000368211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221572.hg.1 | ENST00000412846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221572.hg.1 | ENST00000420938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221572.hg.1 | ENST00000434558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221572.hg.1 | NM_024540.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221572.hg.1 | NM_145729.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221572.hg.1 | TC0100016021.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000361531.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000361531.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000361531.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000361531.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000361531.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000361531.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000361531.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000361531.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000361531.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000368211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000368211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000368211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000368211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000368211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000368211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000368211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000368211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000368211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000412846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000412846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000412846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000412846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000412846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000412846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000420938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000420938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000420938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000434558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000434558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000434558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000434558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000434558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000434558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000478899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000478899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000478899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000478899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | ENST00000478899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | NM_024540.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | NM_024540.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | NM_024540.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | NM_024540.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | NM_024540.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | NM_024540.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | NM_024540.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | NM_024540.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | NM_024540.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | NM_145729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | NM_145729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | NM_145729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | NM_145729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | NM_145729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | NM_145729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | NM_145729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | NM_145729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | NM_145729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016021.hg.1 | NM_145729.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221578.hg.1 | ENST00000368211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221578.hg.1 | NM_024540.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221578.hg.1 | NM_145729.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221578.hg.1 | ENST00000368211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221578.hg.1 | NM_024540.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221578.hg.1 | NM_145729.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221578.hg.1 | TC0100016021.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100221576.hg.1 | ENST00000361531.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221576.hg.1 | ENST00000368211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221576.hg.1 | ENST00000412846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221576.hg.1 | NM_024540.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221576.hg.1 | NM_145729.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221576.hg.1 | ENST00000361531.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221576.hg.1 | ENST00000368211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221576.hg.1 | ENST00000412846.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221576.hg.1 | NM_024540.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221576.hg.1 | NM_145729.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221576.hg.1 | TC0100016021.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 207155 | - | - |

MRPL24
Genomic Map