Assay Details
| Gene Symbol: BMPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204.6 | NP_001195.2 | 6 - 7 | 1998 | 88 | |
| XM_011511687.1 | XP_011509989.1 | 6 - 7 | 2015 | 88 | ||
| GenBank mRNA | AK292430.1 | - | 6 - 7 | 1262 | 88 | |
| BC052985.2 | - | 6 - 7 | 1424 | 6149698 | 88 | |
| D50516.1 | - | 6 - 7 | 1258 | 88 | ||
| U20165.1 | - | 6 - 7 | 886 | 88 | ||
| Z48923.1 | - | 6 - 7 | 1231 | 88 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001192.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
mesoderm formation
maternal placenta development
endothelial cell proliferation
positive regulation of endothelial cell proliferation
lymphangiogenesis
blood vessel remodeling
chondrocyte development
negative regulation of systemic arterial blood pressure
transcription from RNA polymerase II promoter
transmembrane receptor protein serine/threonine kinase signaling pathway
brain development
cellular response to starvation
anterior/posterior pattern specification
positive regulation of endothelial cell migration
positive regulation of epithelial cell migration
positive regulation of pathway-restricted SMAD protein phosphorylation
regulation of lung blood pressure
signal transduction by protein phosphorylation
proteoglycan biosynthetic process
negative regulation of cell growth
positive regulation of bone mineralization
BMP signaling pathway
positive regulation of BMP signaling pathway
activin receptor signaling pathway
regulation of cell proliferation
positive regulation of osteoblast differentiation
negative regulation of vasoconstriction
positive regulation of transcription from RNA polymerase II promoter
vascular endothelial growth factor receptor signaling pathway
lung alveolus development
positive regulation of axon extension involved in axon guidance
limb development
endochondral bone morphogenesis
lymphatic endothelial cell differentiation
artery development
venous blood vessel development
positive regulation of cartilage development
retina vasculature development in camera-type eye
cellular response to BMP stimulus
endothelial cell apoptotic process
negative regulation of chondrocyte proliferation
negative regulation of DNA biosynthetic process
receptor signaling protein serine/threonine kinase activity
protein binding
ATP binding
activin receptor activity, type II
growth factor binding
BMP binding
metal ion binding
BMP receptor activity
mesoderm formation
maternal placenta development
endothelial cell proliferation
positive regulation of endothelial cell proliferation
lymphangiogenesis
blood vessel remodeling
chondrocyte development
negative regulation of systemic arterial blood pressure
transcription from RNA polymerase II promoter
transmembrane receptor protein serine/threonine kinase signaling pathway
brain development
cellular response to starvation
anterior/posterior pattern specification
positive regulation of endothelial cell migration
positive regulation of epithelial cell migration
positive regulation of pathway-restricted SMAD protein phosphorylation
regulation of lung blood pressure
signal transduction by protein phosphorylation
proteoglycan biosynthetic process
negative regulation of cell growth
positive regulation of bone mineralization
BMP signaling pathway
positive regulation of BMP signaling pathway
activin receptor signaling pathway
regulation of cell proliferation
positive regulation of osteoblast differentiation
negative regulation of vasoconstriction
positive regulation of transcription from RNA polymerase II promoter
vascular endothelial growth factor receptor signaling pathway
lung alveolus development
positive regulation of axon extension involved in axon guidance
limb development
endochondral bone morphogenesis
lymphatic endothelial cell differentiation
artery development
venous blood vessel development
positive regulation of cartilage development
retina vasculature development in camera-type eye
cellular response to BMP stimulus
endothelial cell apoptotic process
negative regulation of chondrocyte proliferation
negative regulation of DNA biosynthetic process
receptor signaling protein serine/threonine kinase activity
protein binding
ATP binding
activin receptor activity, type II
growth factor binding
BMP binding
metal ion binding
BMP receptor activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02018140.hg.1 | TC02001192.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02018140.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02018140.hg.1 | ENST00000374580.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02018140.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02018141.hg.1 | TC02001192.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02018141.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02018141.hg.1 | ENST00000374580.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02018141.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000479069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000479069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | ENST00000479069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000479069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000479069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | ENST00000479069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010489.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200174209.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200174209.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200174209.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200174209.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200174209.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200174209.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200174209.hg.1 | TC0200010489.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200174210.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200174210.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200174210.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200174210.hg.1 | ENST00000374574.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200174210.hg.1 | ENST00000374580.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200174210.hg.1 | NM_001204.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200174210.hg.1 | TC0200010489.hg.1 | Clariom D TC |

BMPR2
Genomic Map