Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002836.hg.1 |
ENST00000196169.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002836.hg.1 |
ENST00000377881.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002836.hg.1 |
ENST00000377894.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002836.hg.1 |
ENST00000535286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002836.hg.1 |
NM_001146070.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002836.hg.1 |
NM_001146071.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002836.hg.1 |
NM_030794.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002836.hg.1 |
TC13000240.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300149755.hg.1 |
ENST00000196169.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149755.hg.1 |
ENST00000377881.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149755.hg.1 |
ENST00000377894.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149755.hg.1 |
ENST00000535286.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149755.hg.1 |
ENST00000621840.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149755.hg.1 |
NM_001146070.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149755.hg.1 |
NM_001146071.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149755.hg.1 |
NM_030794.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149755.hg.1 |
ENST00000196169.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149755.hg.1 |
ENST00000377881.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149755.hg.1 |
ENST00000377894.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149755.hg.1 |
ENST00000535286.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149755.hg.1 |
ENST00000621840.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149755.hg.1 |
NM_001146070.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149755.hg.1 |
NM_001146071.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149755.hg.1 |
NM_030794.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149755.hg.1 |
TC1300007344.hg.1 |
Clariom D TC |