Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019829.hg.1 |
TC09001580.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019829.hg.1 |
ENST00000344523.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019829.hg.1 |
ENST00000373584.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019829.hg.1 |
ENST00000416460.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019829.hg.1 |
ENST00000487099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019829.hg.1 |
NM_001489.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019829.hg.1 |
NM_033334.3 |
Refseq |