Assay Details
Gene Symbol: LRRC46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033413.3 | NP_219481.1 | 2 - 3 | 473 | 101 | |
GenBank mRNA | AK292765.1 | - | 2 - 3 | 388 | 101 | |
AK315665.1 | - | 2 - 3 | 473 | 101 | ||
BC010412.2 | - | 2 - 3 | 366 | 3836250 | 101 | |
DB024018.1 | - | 2 - 3 | 473 | 101 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17000614.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17007593.hg.1 | ENST00000269025.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17007593.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17007593.hg.1 | TC17000614.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700159933.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159933.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159933.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159933.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159933.hg.1 | ENST00000584809.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159933.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700159933.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159933.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159933.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159933.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159933.hg.1 | ENST00000584809.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159933.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700159933.hg.1 | TC1700008138.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700159931.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159931.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159931.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159931.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159931.hg.1 | ENST00000584809.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159931.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700159931.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159931.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159931.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159931.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159931.hg.1 | ENST00000584809.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159931.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700159931.hg.1 | TC1700008138.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000269025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579742.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000579971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | ENST00000584809.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008138.hg.1 | NM_033413.3 | Refseq |
