Assay Details
Gene Symbol: CDKAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK310219.1 | - | 5 - 6 | 544 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CDKAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK310219.1 | - | 5 - 6 | 544 | 69 |
Set Membership: |
![]() ![]() ![]() |