Assay Details
| Gene Symbol: PITX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC003685.1 | - | 1 - 2 | 60 | 3452956 | 127 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC003685.1 transcripts.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000265340.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000265340.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000265340.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000265340.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000265340.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000265340.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000265340.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000265340.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000265340.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000265340.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000265340.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000502676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000502676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000502676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000502676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000502676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000502676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000502676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000503586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000503586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000503586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000503586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000503586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000504936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000504936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000504936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000504936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000506438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000506438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000506438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000506438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000506438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000506438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000506438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000506438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000506438.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000507253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000507253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000507253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000507253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000507253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | ENST00000507253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | NM_002653.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | NM_002653.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | NM_002653.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | NM_002653.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | NM_002653.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | NM_002653.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | NM_002653.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | NM_002653.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | NM_002653.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | NM_002653.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012093.hg.1 | NM_002653.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500183672.hg.1 | ENST00000265340.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183672.hg.1 | ENST00000502676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183672.hg.1 | ENST00000506438.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183672.hg.1 | ENST00000507253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183672.hg.1 | NM_002653.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500183672.hg.1 | ENST00000265340.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183672.hg.1 | ENST00000502676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183672.hg.1 | ENST00000506438.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183672.hg.1 | ENST00000507253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183672.hg.1 | NM_002653.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500183672.hg.1 | TC0500012093.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500183678.hg.1 | ENST00000502676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183678.hg.1 | ENST00000506438.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183678.hg.1 | ENST00000502676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183678.hg.1 | ENST00000506438.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183678.hg.1 | TC0500012093.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500183673.hg.1 | ENST00000265340.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183673.hg.1 | ENST00000502676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183673.hg.1 | ENST00000506438.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183673.hg.1 | ENST00000507253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183673.hg.1 | NM_002653.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500183673.hg.1 | ENST00000265340.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183673.hg.1 | ENST00000502676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183673.hg.1 | ENST00000506438.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183673.hg.1 | ENST00000507253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183673.hg.1 | NM_002653.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500183673.hg.1 | TC0500012093.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000265340.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000265340.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000265340.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000265340.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000265340.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000265340.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000265340.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000265340.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000265340.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000265340.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000265340.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000502676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000502676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000502676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000502676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000502676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000502676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000502676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000503586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000503586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000503586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000503586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000503586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000504936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000504936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000504936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000504936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000506438.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000506438.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000506438.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000506438.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000506438.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000506438.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000506438.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000506438.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000506438.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000507253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000507253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000507253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000507253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000507253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | ENST00000507253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | NM_002653.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | NM_002653.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | NM_002653.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | NM_002653.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | NM_002653.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | NM_002653.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | NM_002653.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | NM_002653.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | NM_002653.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | NM_002653.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012093.hg.1 | NM_002653.4 | Refseq |

PITX1
Genomic Map