Assay Details
Gene Symbol: ZNF280C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006724765.3 | XP_006724828.1 | 11 - 12 | 1399 | 67 | |
GenBank mRNA | AL133204.1 | - | 11 - 12 | 1383 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
axon guidance
negative regulation of transcription, DNA-templated
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
metal ion binding
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
axon guidance
negative regulation of transcription, DNA-templated
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
metal ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000447817.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000447817.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000447817.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000447817.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000447817.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000447817.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000447817.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000447817.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000447817.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000447817.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000447817.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000447817.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000447817.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | ENST00000447817.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000370978.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000447817.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000447817.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000447817.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000447817.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000447817.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000447817.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000447817.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000447817.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000447817.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000447817.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000447817.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000447817.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000447817.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | ENST00000447817.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010804.hg.1 | NM_017666.4 | Refseq |
