Assay Details
| Gene Symbol: CLCN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256944.1 | NP_001243873.1 | 7 - 8 | 1565 | 118 | |
| NM_001830.3 | NP_001821.2 | 9 - 10 | 1820 | 118 | ||
| GenBank mRNA | AB019432.1 | - | 8 - 9 | 1665 | 118 | |
| AF170492.1 | - | 7 - 8 | 1390 | 118 | ||
| AK225287.1 | - | 9 - 10 | 1781 | 118 | ||
| AK289564.1 | - | 9 - 10 | 1767 | 118 | ||
| AK299611.1 | - | 6 - 7 | 1198 | 118 | ||
| BC036068.1 | - | 8 - 9 | 1440 | 5311366 | 118 | |
| BC130278.1 | - | 8 - 9 | 1679 | 40146309 | 118 | |
| X77197.1 | - | 9 - 10 | 1773 | 118 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000038.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000454850.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000454850.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000454850.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | ENST00000454850.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000533.hg.1 | TC0X000038.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000533.hg.1 | ENST00000380829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000533.hg.1 | ENST00000380833.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000533.hg.1 | ENST00000421085.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000533.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145076.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145076.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145076.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145076.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145076.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145076.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145076.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145076.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00145076.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145076.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00145076.hg.1 | TC0X00006593.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000380833.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000421085.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000454850.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000454850.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000454850.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | ENST00000454850.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001256944.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006593.hg.1 | NM_001830.3 | Refseq |

CLCN4
Genomic Map