Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009381.hg.1 |
ENST00000402340.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009381.hg.1 |
ENST00000402340.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006924.hg.1 |
ENST00000329357.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006924.hg.1 |
ENST00000329357.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006924.hg.1 |
ENST00000329357.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006924.hg.1 |
NM_152631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006924.hg.1 |
NM_152631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006924.hg.1 |
NM_152631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001828.hg.1 |
TC0X000159.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001828.hg.1 |
ENST00000329357.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001828.hg.1 |
NM_152631.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006924.hg.1 |
ENST00000329357.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006924.hg.1 |
ENST00000329357.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006924.hg.1 |
ENST00000329357.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006924.hg.1 |
NM_152631.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006924.hg.1 |
NM_152631.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006924.hg.1 |
NM_152631.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009381.hg.1 |
ENST00000402340.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009381.hg.1 |
ENST00000402340.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147157.hg.1 |
ENST00000329357.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147157.hg.1 |
NM_152631.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147157.hg.1 |
ENST00000329357.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147157.hg.1 |
NM_152631.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147157.hg.1 |
TC0X00006924.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00147156.hg.1 |
ENST00000329357.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147156.hg.1 |
NM_152631.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147156.hg.1 |
ENST00000329357.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147156.hg.1 |
NM_152631.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147156.hg.1 |
TC0X00006924.hg.1 |
Clariom D TC |