Assay Details
| Gene Symbol: GLP2R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004246.2 | NP_004237.1 | 3 - 4 | 898 | 69 | |
| GenBank mRNA | AF105367.1 | - | 3 - 4 | 385 | 69 | |
| AK295765.1 | - | 3 - 4 | 277 | 69 | ||
| AK307283.1 | - | 4 - 5 | 791 | 69 | ||
| AK314269.1 | - | 3 - 4 | 898 | 69 | ||
| BC043390.1 | - | 1 - 2 | 93 | 69 | ||
| BC096261.1 | - | 3 - 4 | 385 | 69 | ||
| BC096262.1 | - | 3 - 4 | 385 | 40006110 | 69 | |
| BC096263.1 | - | 3 - 4 | 381 | 69 | ||
| BC096264.1 | - | 3 - 4 | 385 | 69 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000147.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17002042.hg.1 | ENST00000262441.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002042.hg.1 | ENST00000304773.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002042.hg.1 | ENST00000396206.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002042.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002042.hg.1 | TC17000147.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000304773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000304773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000304773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000304773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000304773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000304773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000304773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000304773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148353.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148353.hg.1 | ENST00000304773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148353.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148353.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148353.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148353.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148353.hg.1 | ENST00000304773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148353.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148353.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148353.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148353.hg.1 | TC1700006860.hg.1 | Clariom D TC |

GLP2R
Genomic Map