Assay Details
| Gene Symbol: RGS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005273218.3 | XP_005273275.1 | 17 - 18 | 1940 | 102 | |
| XM_006711800.3 | XP_006711863.1 | 16 - 17 | 1731 | 102 | ||
| XM_011544246.2 | XP_011542548.1 | 16 - 17 | 1390 | 102 | ||
| XM_011544247.2 | XP_011542549.1 | 16 - 17 | 1435 | 102 | ||
| XM_017002001.1 | XP_016857490.1 | 16 - 17 | 1889 | 102 | ||
| XM_017002002.1 | XP_016857491.1 | 18 - 19 | 1815 | 102 | ||
| XM_017002011.1 | XP_016857500.1 | 14 - 15 | 2902 | 102 | ||
| GenBank mRNA | AF090117.1 | - | 10 - 11 | 907 | 102 | |
More Information
Set Membership: |
|

RGS7
Genomic Map