Assay Details
| Gene Symbol: LMF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033200.2 | NP_149977.2 | 4 - 5 | 630 | 84 | |
| XM_005261955.3 | XP_005262012.1 | 4 - 5 | 712 | 84 | ||
| XM_017029077.1 | XP_016884566.1 | 4 - 5 | 764 | 84 | ||
| GenBank mRNA | BC002942.2 | - | 4 - 5 | 604 | 3954961 | 84 |
| BC014652.1 | - | 4 - 5 | 693 | 4562676 | 84 | |
| BC021143.1 | - | 4 - 5 | 604 | 4872642 | 84 | |
| CR456605.1 | - | 4 - 5 | 614 | 84 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000914.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22015598.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015598.hg.1 | ENST00000380796.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015598.hg.1 | ENST00000474879.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015598.hg.1 | ENST00000505981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015598.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015598.hg.1 | TC22000914.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000507607.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000507607.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000507607.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000507607.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000507607.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015600.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015600.hg.1 | ENST00000380796.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015600.hg.1 | ENST00000474879.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015600.hg.1 | ENST00000505981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015600.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015600.hg.1 | TC22000914.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2200166504.hg.1 | TC2200009161.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200166504.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166504.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166504.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166504.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166504.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200166504.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166504.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166504.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166504.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166504.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000507607.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000507607.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000507607.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000507607.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000507607.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200166506.hg.1 | TC2200009161.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200166506.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166506.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166506.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166506.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200166506.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166506.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166506.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166506.hg.1 | NM_033200.2 | Refseq |

LMF2
Genomic Map