Assay Details
| Gene Symbol: DPY30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032574.3 | NP_115963.1 | 1 - 2 | 244 | 75 | |
| XM_011533133.2 | XP_011531435.1 | 1 - 2 | 177 | 75 | ||
| GenBank mRNA | AF226998.1 | - | 1 - 2 | 163 | 75 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02005024.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF226998.1,NM_032574.3,XM_011533133.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
chromatin silencing at telomere
transcription, DNA-templated
endosomal transport
histone H3-K4 methylation
protein binding
histone-lysine N-methyltransferase activity
histone methyltransferase activity (H3-K4 specific)
identical protein binding
protein homodimerization activity
chromatin silencing at telomere
transcription, DNA-templated
endosomal transport
histone H3-K4 methylation
protein binding
histone-lysine N-methyltransferase activity
histone methyltransferase activity (H3-K4 specific)
identical protein binding
protein homodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02027765.hg.1 | TC02005024.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027765.hg.1 | ENST00000295066.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027765.hg.1 | ENST00000342166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027765.hg.1 | NM_032574.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027764.hg.1 | TC02005024.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027764.hg.1 | ENST00000295066.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027764.hg.1 | ENST00000342166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027764.hg.1 | NM_032574.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027766.hg.1 | TC02005024.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027766.hg.1 | ENST00000295066.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027766.hg.1 | NM_032574.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000295066.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000295066.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000295066.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000295066.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000295066.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000295066.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000295066.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000342166.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000342166.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000342166.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000342166.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000342166.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000342166.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000342166.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000414013.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000414013.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000414013.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000414013.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000414013.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000414013.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000422413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000422413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000422413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000422413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000422413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000422413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000422413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000430665.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000430665.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000430665.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000430665.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000430665.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000430665.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000430665.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000452582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000452582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000452582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000452582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000452582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000452582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | ENST00000452582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | NM_032574.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | NM_032574.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | NM_032574.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | NM_032574.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | NM_032574.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | NM_032574.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | NM_032574.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016647.hg.1 | NM_032574.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200226017.hg.1 | ENST00000295066.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226017.hg.1 | ENST00000342166.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226017.hg.1 | ENST00000414013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226017.hg.1 | ENST00000422413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226017.hg.1 | ENST00000430665.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226017.hg.1 | ENST00000452582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226017.hg.1 | NM_032574.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200226017.hg.1 | ENST00000295066.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226017.hg.1 | ENST00000342166.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226017.hg.1 | ENST00000414013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226017.hg.1 | ENST00000422413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226017.hg.1 | ENST00000430665.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226017.hg.1 | ENST00000452582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226017.hg.1 | NM_032574.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200226017.hg.1 | TC0200016647.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200226019.hg.1 | ENST00000295066.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226019.hg.1 | NM_032574.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200226019.hg.1 | ENST00000295066.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226019.hg.1 | NM_032574.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200226019.hg.1 | TC0200016647.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000295066.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000295066.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000295066.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000295066.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000295066.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000295066.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000295066.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000342166.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000342166.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000342166.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000342166.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000342166.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000342166.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000342166.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000414013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000414013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000414013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000414013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000414013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000414013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000422413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000422413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000422413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000422413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000422413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000422413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000422413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000430665.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000430665.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000430665.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000430665.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000430665.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000430665.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000430665.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000452582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000452582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000452582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000452582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000452582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000452582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | ENST00000452582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | NM_032574.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | NM_032574.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | NM_032574.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | NM_032574.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | NM_032574.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | NM_032574.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | NM_032574.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016647.hg.1 | NM_032574.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200226016.hg.1 | ENST00000295066.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226016.hg.1 | ENST00000342166.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226016.hg.1 | ENST00000414013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226016.hg.1 | ENST00000422413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226016.hg.1 | ENST00000430665.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226016.hg.1 | ENST00000452582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226016.hg.1 | NM_032574.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200226016.hg.1 | ENST00000295066.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226016.hg.1 | ENST00000342166.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226016.hg.1 | ENST00000414013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226016.hg.1 | ENST00000422413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226016.hg.1 | ENST00000430665.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226016.hg.1 | ENST00000452582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226016.hg.1 | NM_032574.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200226016.hg.1 | TC0200016647.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200226018.hg.1 | ENST00000295066.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226018.hg.1 | ENST00000342166.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226018.hg.1 | ENST00000414013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226018.hg.1 | ENST00000422413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226018.hg.1 | ENST00000430665.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226018.hg.1 | ENST00000452582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226018.hg.1 | NM_032574.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200226018.hg.1 | ENST00000295066.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226018.hg.1 | ENST00000342166.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226018.hg.1 | ENST00000414013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226018.hg.1 | ENST00000422413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226018.hg.1 | ENST00000430665.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226018.hg.1 | ENST00000452582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200226018.hg.1 | NM_032574.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200226018.hg.1 | TC0200016647.hg.1 | Clariom D TC |

DPY30
Genomic Map