Assay Details
| Gene Symbol: TPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004179.2 | NP_004170.1 | 7 - 8 | 956 | 95 | |
| GenBank mRNA | BC106739.1 | - | 7 - 8 | 930 | 40016887 | 95 |
| BC106740.2 | - | 6 - 7 | 733 | 95 | ||
| X52836.1 | - | 7 - 8 | 930 | 95 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11003436.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
circadian rhythm
aromatic amino acid family metabolic process
negative regulation of ossification
response to immobilization stress
serotonin biosynthetic process
positive regulation of fat cell differentiation
indolalkylamine biosynthetic process
bone remodeling
oxidation-reduction process
mammary gland alveolus development
tryptophan 5-monooxygenase activity
iron ion binding
amino acid binding
circadian rhythm
aromatic amino acid family metabolic process
negative regulation of ossification
response to immobilization stress
serotonin biosynthetic process
positive regulation of fat cell differentiation
indolalkylamine biosynthetic process
bone remodeling
oxidation-reduction process
mammary gland alveolus development
tryptophan 5-monooxygenase activity
iron ion binding
amino acid binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11018436.hg.1 | TC11003436.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018436.hg.1 | ENST00000250018.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018436.hg.1 | ENST00000341556.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018436.hg.1 | NM_004179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000417164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000417164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000417164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000417164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000417164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000417164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000417164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000417164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000417164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000417164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000525406.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000525406.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000525406.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000528338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000528338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000528338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000528338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000528338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | ENST00000528338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | NM_004179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | NM_004179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | NM_004179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | NM_004179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | NM_004179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | NM_004179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | NM_004179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | NM_004179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | NM_004179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | NM_004179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | NM_004179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | NM_004179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013150.hg.1 | NM_004179.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203828.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203828.hg.1 | ENST00000417164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203828.hg.1 | NM_004179.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203828.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203828.hg.1 | ENST00000417164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203828.hg.1 | NM_004179.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203828.hg.1 | TC1100013150.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000250018.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000417164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000417164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000417164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000417164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000417164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000417164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000417164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000417164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000417164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000417164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000525406.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000525406.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000525406.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000528338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000528338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000528338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000528338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000528338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | ENST00000528338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | NM_004179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | NM_004179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | NM_004179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | NM_004179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | NM_004179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | NM_004179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | NM_004179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | NM_004179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | NM_004179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | NM_004179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | NM_004179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | NM_004179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013150.hg.1 | NM_004179.2 | Refseq |

TPH1
Genomic Map