Assay Details
| Gene Symbol: DNAH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006721062.1 | XP_006721125.1 | 2 - 3 | 309 | 62 | |
| GenBank mRNA | AK056509.2 | - | 2 - 3 | 309 | 62 | |
| AK303958.1 | - | 2 - 3 | 318 | 62 | ||
| AK310973.1 | - | 2 - 3 | 306 | 62 | ||
More Information
Set Membership: |
|

DNAH3
Genomic Map