Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014665.hg.1 |
TC08001142.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014665.hg.1 |
ENST00000311351.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014665.hg.1 |
ENST00000520755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014665.hg.1 |
ENST00000522515.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014665.hg.1 |
NM_014462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014665.hg.1 |
NR_045492.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014667.hg.1 |
TC08001142.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014667.hg.1 |
ENST00000311351.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014667.hg.1 |
ENST00000520755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014667.hg.1 |
ENST00000522515.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014667.hg.1 |
NM_014462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014667.hg.1 |
NR_045492.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014666.hg.1 |
TC08001142.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014666.hg.1 |
ENST00000311351.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014666.hg.1 |
ENST00000520755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014666.hg.1 |
ENST00000522515.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014666.hg.1 |
NM_014462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014666.hg.1 |
NR_045492.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014663.hg.1 |
TC08001142.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014663.hg.1 |
ENST00000311351.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014663.hg.1 |
ENST00000520755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014663.hg.1 |
ENST00000522515.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014663.hg.1 |
NM_014462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014663.hg.1 |
NR_045492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800169030.hg.1 |
ENST00000311351.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169030.hg.1 |
ENST00000520755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169030.hg.1 |
ENST00000522515.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169030.hg.1 |
ENST00000523511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169030.hg.1 |
NM_014462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800169030.hg.1 |
NR_045492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800169030.hg.1 |
ENST00000311351.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169030.hg.1 |
ENST00000520755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169030.hg.1 |
ENST00000522515.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169030.hg.1 |
ENST00000523511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169030.hg.1 |
NM_014462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800169030.hg.1 |
NR_045492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800169030.hg.1 |
TC0800010154.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800169029.hg.1 |
ENST00000311351.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169029.hg.1 |
ENST00000520755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169029.hg.1 |
ENST00000522515.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169029.hg.1 |
ENST00000523511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169029.hg.1 |
NM_014462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800169029.hg.1 |
NR_045492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800169029.hg.1 |
ENST00000311351.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169029.hg.1 |
ENST00000520755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169029.hg.1 |
ENST00000522515.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169029.hg.1 |
ENST00000523511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169029.hg.1 |
NM_014462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800169029.hg.1 |
NR_045492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800169029.hg.1 |
TC0800010154.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800169031.hg.1 |
ENST00000311351.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169031.hg.1 |
ENST00000520755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169031.hg.1 |
ENST00000522515.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169031.hg.1 |
ENST00000523511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169031.hg.1 |
NM_014462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800169031.hg.1 |
NR_045492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800169031.hg.1 |
ENST00000311351.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169031.hg.1 |
ENST00000520755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169031.hg.1 |
ENST00000522515.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169031.hg.1 |
ENST00000523511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169031.hg.1 |
NM_014462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800169031.hg.1 |
NR_045492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800169031.hg.1 |
TC0800010154.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800169032.hg.1 |
ENST00000311351.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169032.hg.1 |
ENST00000520755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169032.hg.1 |
ENST00000522515.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169032.hg.1 |
NM_014462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800169032.hg.1 |
NR_045492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800169032.hg.1 |
ENST00000311351.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169032.hg.1 |
ENST00000520755.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169032.hg.1 |
ENST00000522515.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169032.hg.1 |
NM_014462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800169032.hg.1 |
NR_045492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800169032.hg.1 |
TC0800010154.hg.1 |
Clariom D TC |