Assay Details
Gene Symbol: LSM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014462.2 | NP_055277.1 | 2 - 3 | 528 | 98 | |
NR_045493.1 | 2 - 3 | 250 | 98 | |||
GenBank mRNA | AF000177.1 | - | 2 - 3 | 282 | 98 | |
AF147348.1 | - | 2 - 3 | 171 | 98 | ||
AJ238094.1 | - | 2 - 3 | 306 | 98 | ||
AK312159.1 | - | 2 - 3 | 281 | 98 | ||
BC001767.1 | - | 2 - 3 | 215 | 3353833 | 98 | |
BP376421.1 | - | 2 - 3 | 491 | 98 | ||
BU941962.1 | - | 2 - 3 | 253 | 98 | ||
KU178606.1 | - | 2 - 3 | 118 | 98 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08001142.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
deadenylation-dependent decapping of nuclear-transcribed mRNA
RNA splicing, via transesterification reactions
mRNA processing
RNA splicing
stem cell population maintenance
exonucleolytic nuclear-transcribed mRNA catabolic process involved in deadenylation-dependent decay
negative regulation of neuron differentiation
histone mRNA catabolic process
RNA cap binding
mRNA binding
protein binding
pre-mRNA binding
poly(A) RNA binding
deadenylation-dependent decapping of nuclear-transcribed mRNA
RNA splicing, via transesterification reactions
mRNA processing
RNA splicing
stem cell population maintenance
exonucleolytic nuclear-transcribed mRNA catabolic process involved in deadenylation-dependent decay
negative regulation of neuron differentiation
histone mRNA catabolic process
RNA cap binding
mRNA binding
protein binding
pre-mRNA binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR08014659.hg.1 | TC08001142.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08014659.hg.1 | ENST00000311351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08014659.hg.1 | ENST00000522515.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08014659.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08014659.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800169025.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800169025.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800169025.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800169025.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800169025.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800169025.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800169025.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800169025.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800169025.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800169025.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800169025.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800169025.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800169025.hg.1 | TC0800010154.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000311351.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520286.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000520755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000522515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | ENST00000523511.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NM_014462.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045492.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045493.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010154.hg.1 | NR_045493.1 | Refseq |
