Assay Details
| Gene Symbol: LOC100499484 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036526.1 | 13 - 14 | 1206 | 87 | ||
| GenBank mRNA | AK301827.1 | - | 13 - 14 | 1149 | 87 | |
More Information
Set Membership: |
|

LOC100499484
Genomic Map