Assay Details
Gene Symbol: TRIM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF220131.1 | - | 4 - 5 | 699 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
